siRNA Details
| Gene Symbol: CLCA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012128.3 | NP_036260.2 | 10 | 1531 | ||
| NR_024602.1 | 9 | 1422 | ||||
| XM_011541015.2 | XP_011539317.1 | 10 | 1350 | |||
| GenBank mRNA | AF127035.1 | - | 10 | 1515 | ||
| AK000072.1 | - | 10 | 1529 | |||
| AK000138.1 | - | 3 | 208 | |||
| AK307785.1 | - | 9 | 1367 | |||
| AK307898.1 | - | 8 | 1136 | |||
| AK314430.1 | - | 10 | 1527 | |||
| AY358470.1 | - | 10 | 1511 | |||
| BC113687.1 | - | 10 | 1526 | 8322739 | ||
| BC113689.1 | - | 10 | 1525 | 8322741 | ||

CLCA4
Genomic Map