siRNA Details
Gene Symbol: Chst5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019950.2 | NP_064334.1 | 2 | 616 | ||
GenBank mRNA | AF176840.1 | - | 2 | 639 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Chst5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019950.2 | NP_064334.1 | 2 | 616 | ||
GenBank mRNA | AF176840.1 | - | 2 | 639 |