siRNA Details
| Gene Symbol: Pdgfc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019971.2 | NP_064355.1 | 3 | 1420 | ||
| GenBank mRNA | AF117608.1 | - | 3 | 596 | ||
| AF266467.1 | - | 3 | 1420 | |||
| AF286725.1 | - | 3 | 399 | |||
| AK033734.1 | - | 3 | 1149 | |||
| AK042767.1 | - | 3 | 1167 | |||
| AK052947.1 | - | 3 | 672 | |||
| AK081347.1 | - | 4 | 472 | |||
| BC029099.1 | - | 4 | 723 | |||

Pdgfc
Genomic Map