siRNA Details
| Gene Symbol: Stap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019992.3 | NP_064376.1 | 2 | 169 | ||
| GenBank mRNA | AB036058.1 | - | 2 | 159 | ||
| AK041474.1 | - | 2 | 170 | |||
| AK042475.1 | - | 2 | 216 | |||
| AK153661.1 | - | 2 | 248 | |||
| AK162655.1 | - | 2 | 251 | |||
| BC057898.1 | - | 2 | 223 | |||
| BC145550.1 | - | 2 | 185 | 9054114 | ||
| BC145828.1 | - | 2 | 184 | 40131131 | ||
| BC145830.1 | - | 2 | 185 | 40131133 | ||

Stap1
Genomic Map