siRNA Details
| Gene Symbol: Sult1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019878.4 | NP_063931.1 | 8 | 1164 | ||
| GenBank mRNA | AK033498.1 | - | 8 | 1167 | ||
| BC024361.1 | - | 7 | 1123 | 4990345 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sult1b1| Gene Symbol: Sult1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019878.4 | NP_063931.1 | 8 | 1164 | ||
| GenBank mRNA | AK033498.1 | - | 8 | 1167 | ||
| BC024361.1 | - | 7 | 1123 | 4990345 | ||