siRNA Details
| Gene Symbol: Crtap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019922.2 | NP_064306.2 | 3 | 889 | ||
| GenBank mRNA | AJ006469.1 | - | 3 | 847 | ||
| AK017797.1 | - | 3 | 852 | |||
| AK047506.1 | - | 3 | 845 | |||
| AK203540.1 | - | 1 | 29 | |||
| BC049890.1 | - | 3 | 826 | 6306169 | ||

Crtap
Genomic Map