siRNA Details
| Gene Symbol: Ovol1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019935.3 | NP_064319.1 | 4 | 2222 | ||
| GenBank mRNA | AF134804.1 | - | 4 | 2238 | ||
| AK028577.1 | - | 4 | 2223 | |||
| AK132478.1 | - | 4 | 2224 | |||
| AK132505.1 | - | 4 | 2262 | |||
| AK132506.1 | - | 4 | 2257 | |||
| AK144952.1 | - | 4 | 2259 | |||

Ovol1
Genomic Map