siRNA Details
| Gene Symbol: Cript | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019936.3 | NP_064320.1 | 5 | 374 | ||
| GenBank mRNA | AF151637.1 | - | 5 | 339 | ||
| AK004828.1 | - | 5 | 367 | |||
| AK008523.1 | - | 5 | 359 | |||
| AK029409.1 | - | 5 | 362 | |||
| AK213321.1 | - | 3 | 206 | |||
| BC002258.1 | - | 4 | 256 | |||
| BC023013.1 | - | 5 | 348 | 5369938 | ||

Cript
Genomic Map