siRNA Details
| Gene Symbol: Zfp111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019940.2 | NP_064324.2 | 4 | 2759 | ||
| GenBank mRNA | AF167318.1 | - | 2 | 2666 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp111| Gene Symbol: Zfp111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019940.2 | NP_064324.2 | 4 | 2759 | ||
| GenBank mRNA | AF167318.1 | - | 2 | 2666 | ||