siRNA Details
| Gene Symbol: Zfp235 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019941.2 | NP_064325.2 | 4 | 3369 | ||
| GenBank mRNA | AF167321.1 | - | 4 | 3330 | ||
| BC099965.1 | - | 4 | 3289 | 30084081 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp235| Gene Symbol: Zfp235 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019941.2 | NP_064325.2 | 4 | 3369 | ||
| GenBank mRNA | AF167321.1 | - | 4 | 3330 | ||
| BC099965.1 | - | 4 | 3289 | 30084081 | ||