siRNA Details
| Gene Symbol: Papolb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019943.2 | NP_064327.1 | 1 | 1491 | ||
| GenBank mRNA | AB030729.1 | - | 1 | 1491 | ||
| AB221598.1 | - | 1 | 1301 | |||
| AF039957.1 | - | 1 | 1440 | |||
| BC129896.1 | - | 1 | 1469 | 40111521 | ||
| BC129897.1 | - | 1 | 1469 | 40111523 | ||

Papolb
Genomic Map