siRNA Details
| Gene Symbol: KCNK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017424.2 | NP_001017424.1 | 2 | 651 | ||
| NM_001017425.2 | NP_001017425.2 | 2 | 452 | |||
| NM_014217.3 | NP_055032.1 | 2 | 328 | |||
| XM_011509522.2 | XP_011507824.1 | 2 | 1854 | |||
| XM_011509523.1 | XP_011507825.1 | 2 | 256 | |||
| XM_011509524.2 | XP_011507826.1 | 2 | 303 | |||
| XM_017001248.1 | XP_016856737.1 | 3 | 1039 | |||
| XM_017001249.1 | XP_016856738.1 | 3 | 990 | |||
| GenBank mRNA | AF004711.1 | - | 2 | 436 | ||
| AF129399.1 | - | 1 | 238 | |||
| AF171068.1 | - | 2 | 252 | |||
| AK123409.1 | - | 2 | 340 | |||
| AK291483.1 | - | 2 | 432 | |||
| AK294780.1 | - | 2 | 434 | |||
| AK315249.1 | - | 3 | 418 | |||
| AU280191.1 | - | 2 | 394 | |||
| AY552980.2 | - | 2 | 338 | |||
| AY552981.1 | - | 2 | 343 | |||
| BC069462.1 | - | 2 | 252 | 7262344 | ||
| BC101693.1 | - | 2 | 297 | 8069199 | ||
| BC101695.1 | - | 2 | 297 | 8069201 | ||
| BC143586.1 | - | 2 | 298 | 9052097 | ||
| EF165334.1 | - | 2 | 385 | |||
| EF165335.1 | - | 2 | 350 | |||
| KF182338.1 | - | 2 | 287 | |||
| KF182339.1 | - | 2 | 261 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001281848, NM_001159850, NM_001281847, NM_010607, U73488, and others.

KCNK2
Genomic Map