siRNA Details
| Gene Symbol: Cnpy2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019953.1 | NP_064337.1 | 5 | 785 | ||
| GenBank mRNA | AF186115.1 | - | 5 | 785 | ||
| AK007914.1 | - | 5 | 546 | |||
| AK013014.1 | - | 5 | 656 | |||
| AK013568.1 | - | 5 | 527 | |||
| AK019927.1 | - | 5 | 657 | |||
| AK150953.1 | - | 5 | 729 | |||
| BC008261.1 | - | 5 | 557 | 2650612 | ||

Cnpy2
Genomic Map