siRNA Details
| Gene Symbol: C1qtnf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204129.1 | NP_001191058.1 | 4 | 1513 | ||
| NM_001204130.1 | NP_001191059.1 | 4 | 1425 | |||
| NM_019959.3 | NP_064343.1 | 4 | 1503 | |||
| GenBank mRNA | AF192499.1 | - | 4 | 1450 | ||
| AK005484.1 | - | 4 | 1472 | |||
| BC023468.1 | - | 1 | 1102 | |||
| BC037636.1 | - | 3 | 1416 | |||
| BC049894.1 | - | 4 | 1452 | 4018549 | ||

C1qtnf1
Genomic Map