siRNA Details
| Gene Symbol: Pdgfc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019971.2 | NP_064355.1 | 3 | 1451 | ||
| GenBank mRNA | AF117608.1 | - | 3 | 627 | ||
| AF266467.1 | - | 3 | 1451 | |||
| AF286725.1 | - | 3 | 430 | |||
| AK033734.1 | - | 3 | 1180 | |||
| AK042767.1 | - | 3 | 1198 | |||
| AK052947.1 | - | 3 | 703 | |||
| AK081347.1 | - | 4 | 503 | |||
| BC029099.1 | - | 4 | 754 | |||
| BC037696.1 | - | 3 | 638 | 4008749 | ||

Pdgfc
Genomic Map