siRNA Details
| Gene Symbol: Sh3bgrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019989.2 | NP_064373.1 | 4 | 842 | ||
| GenBank mRNA | AJ272504.1 | - | 4 | 712 | ||
| AK049929.1 | - | 4 | 734 | |||
| AK146588.1 | - | 4 | 750 | |||
| AK150604.1 | - | 4 | 750 | |||
| AK151095.1 | - | 4 | 752 | |||
| AK151501.1 | - | 4 | 712 | |||
| AK151871.1 | - | 4 | 754 | |||
| AK165995.1 | - | 4 | 752 | |||
| BC034161.1 | - | 4 | 796 | 4954986 | ||
| BC043055.1 | - | 4 | 842 | 6418767 | ||

Sh3bgrl
Genomic Map