siRNA Details
| Gene Symbol: Alg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019998.3 | NP_064382.3 | 2 | 796 | ||
| GenBank mRNA | AB041604.1 | - | 2 | 733 | ||
| AB161357.1 | - | 2 | 730 | |||
| AK004997.2 | - | 2 | 754 | |||
| AK010673.1 | - | 2 | 762 | |||
| AK181454.1 | - | 1 | 201 | |||
| BC051951.1 | - | 2 | 744 | 5686826 | ||
| BC052411.1 | - | 2 | 749 | 5720077 | ||

Alg2
Genomic Map