siRNA Details
| Gene Symbol: Med8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290688.1 | NP_001277617.1 | 6, 7 | 754 | ||
| NM_020000.3 | NP_064384.2 | 6, 7 | 794 | |||
| NM_173719.3 | NP_776067.1 | 7, 8 | 692 | |||
| GenBank mRNA | AB041805.1 | - | 5, 6 | 2280 | ||
| AK005422.1 | - | 7, 8 | 675 | |||
| AK008759.1 | - | 6, 7 | 773 | |||
| AK011080.1 | - | 6, 7 | 754 | |||
| AK147032.1 | - | 6, 7 | 780 | |||
| AK148025.1 | - | 6, 7 | 775 | |||
| BC002315.1 | - | 5, 6 | 734 | |||
| BC021870.1 | - | 6, 7 | 765 | 4022630 | ||
| BC051683.1 | - | 6, 7 | 732 | |||
| BC053738.1 | - | 4, 5 | 1641 | |||

Med8
Genomic Map