siRNA Details
| Gene Symbol: Rec8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020002.3 | NP_064386.2 | 11 | 1100 | ||
| GenBank mRNA | AF262055.1 | - | 11 | 1030 | ||
| AK077167.1 | - | 11 | 1035 | |||
| AK146832.1 | - | 11 | 1086 | |||
| BC052155.1 | - | 11 | 1096 | 6335959 | ||

Rec8
Genomic Map