siRNA Details
| Gene Symbol: Pfdn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027044.3 | NP_081320.2 | 2 | 192 | ||
| GenBank mRNA | AF108357.1 | - | 2 | 152 | ||
| AK004438.1 | - | 2 | 147 | |||
| AK005682.1 | - | 2 | 138 | |||
| AK028206.1 | - | 2 | 129 | |||
| AK145883.1 | - | 2 | 145 | |||
| BC026920.1 | - | 2 | 129 | 3666296 | ||
| CN835522.1 | - | 2 | 195 | |||

Pfdn5
Genomic Map