siRNA Details
| Gene Symbol: Tmem9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020050.1 | NP_064434.1 | 5 | 710 | ||
| GenBank mRNA | AK007430.1 | - | 5 | 678 | ||
| AK009147.1 | - | 5 | 645 | |||
| AK049882.1 | - | 5 | 797 | |||
| AK077253.1 | - | 5 | 666 | |||
| AK147127.1 | - | 5 | 726 | |||
| AK207353.1 | - | 1 | 41 | |||
| AK215177.1 | - | 1 | 41 | |||
| BC002208.1 | - | 5 | 694 | 3490055 | ||

Tmem9b
Genomic Map