siRNA Details
| Gene Symbol: Clec2i | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020257.2 | NP_064653.1 | 2, 3 | 159 | ||
| GenBank mRNA | AF121352.1 | - | 2, 3 | 132 | ||
| AF350411.1 | - | 2, 3 | 136 | |||
| AY137345.1 | - | 2, 3 | 158 | |||
| BC139192.1 | - | 2, 3 | 132 | 8862214 | ||
| BC139193.1 | - | 2, 3 | 132 | 8862215 | ||
| HQ713440.1 | - | 1, 2 | 66 | |||

Clec2i
Genomic Map