siRNA Details
Gene Symbol: Psg23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020261.4 | NP_064657.2 | 2 | 423 | ||
GenBank mRNA | AF113599.1 | - | 238 | |||
AK018828.1 | - | 2 | 424 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Psg23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020261.4 | NP_064657.2 | 2 | 423 | ||
GenBank mRNA | AF113599.1 | - | 238 | |||
AK018828.1 | - | 2 | 424 |