siRNA Details
| Gene Symbol: Trim44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020267.2 | NP_064663.2 | 1 | 536 | ||
| GenBank mRNA | AJ249129.1 | - | 1 | 482 | ||
| AJ278191.1 | - | 1 | 544 | |||
| BC098369.1 | - | 1 | 505 | 6830438 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim44| Gene Symbol: Trim44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020267.2 | NP_064663.2 | 1 | 536 | ||
| GenBank mRNA | AJ249129.1 | - | 1 | 482 | ||
| AJ278191.1 | - | 1 | 544 | |||
| BC098369.1 | - | 1 | 505 | 6830438 | ||