siRNA Details
| Gene Symbol: Klk1b27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020268.3 | NP_064664.1 | 2 | 125 | ||
| GenBank mRNA | AB039275.1 | - | 2 | 127 | ||
| BC132090.1 | - | 2 | 102 | 40130367 | ||
| BC132665.1 | - | 2 | 102 | 40130942 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klk1b27| Gene Symbol: Klk1b27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020268.3 | NP_064664.1 | 2 | 125 | ||
| GenBank mRNA | AB039275.1 | - | 2 | 127 | ||
| BC132090.1 | - | 2 | 102 | 40130367 | ||
| BC132665.1 | - | 2 | 102 | 40130942 | ||