siRNA Details
| Gene Symbol: Tob2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020507.3 | NP_065253.1 | 2 | 799 | ||
| GenBank mRNA | AK049766.1 | - | 2 | 746 | ||
| AK122537.1 | - | 1 | 2004 | |||
| AK143542.1 | - | 2 | 776 | |||
| BC011163.1 | - | 1 | 681 | 4194136 | ||
| BC021869.1 | - | 2 | 733 | |||
| BC094031.1 | - | 2 | 730 | 4945656 | ||

Tob2
Genomic Map