siRNA Details
| Gene Symbol: Pigo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB038560.1 | - | 1 | 1017 | ||
| AK050819.1 | - | 2 | 356 | |||
| AK220315.1 | - | 1 | 965 | |||
| BC036977.1 | - | 2 | 362 | |||
| BC092394.1 | - | 1 | 1035 | 30534262 | ||
| BC141058.1 | - | 1 | 973 | 9055670 | ||

Pigo
Genomic Map