siRNA Details
| Gene Symbol: Igdcc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290315.1 | NP_001277244.1 | 2 | 273 | ||
| NM_020043.3 | NP_064427.2 | 2 | 273 | |||
| GenBank mRNA | AB052620.1 | - | 2 | 273 | ||
| AB052621.1 | - | 2 | 273 | |||
| AF176694.1 | - | 2 | 224 | |||
| AK079073.1 | - | 2 | 238 | |||
| AK082162.1 | - | 2 | 267 | |||
| AK122535.1 | - | 202 | ||||
| AK143256.1 | - | 2 | 226 | |||
| BB624536.1 | - | 2 | 264 | |||
| BC126880.1 | - | 2 | 224 | |||

Igdcc4
Genomic Map