siRNA Details
| Gene Symbol: Rbms1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001141931.1 | NP_001135403.1 | 3 | 545 | ||
| NM_001141932.1 | NP_001135404.1 | 3 | 545 | |||
| NM_020296.2 | NP_064692.2 | 3 | 738 | |||
| GenBank mRNA | AB026569.1 | - | 3 | 461 | ||
| AK133054.1 | - | 3 | 266 | |||
| AK135280.1 | - | 3 | 425 | |||
| AK147109.1 | - | 3 | 739 | |||
| AK161102.1 | - | 3 | 495 | |||
| AK164307.1 | - | 3 | 539 | |||
| BC016501.1 | - | 3 | 726 | 4527742 | ||
| BC057866.1 | - | 3 | 577 | 5327143 | ||

Rbms1
Genomic Map