siRNA Details
| Gene Symbol: Dolpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290508.1 | NP_001277437.1 | 7 | 950 | ||
| NM_020329.4 | NP_065062.1 | 6 | 743 | |||
| GenBank mRNA | AB030189.1 | - | 6 | 583 | ||
| AK002544.1 | - | 7 | 789 | |||
| AK010797.1 | - | 2 | 180 | |||
| AK044141.1 | - | 6 | 582 | |||
| AK053923.1 | - | 6 | 572 | |||
| AK088065.1 | - | 6 | 586 | |||
| BC026544.1 | - | 6 | 567 | 4975127 | ||

Dolpp1
Genomic Map