siRNA Details
| Gene Symbol: Magea2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020016.1 | NP_064400.1 | 4 | 1481 | ||
| Gene Symbol: Magea1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020015.2 | NP_064399.1 | 1 | 1244 | ||
| GenBank mRNA | CJ223782.1 | - | 1 | 359 | ||

Magea2
Genomic Map