siRNA Details
Gene Symbol: Hist1h1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020034.2 | NP_064418.1 | 1 | 730 | ||
GenBank mRNA | BC114579.1 | - | 1 | 658 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hist1h1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020034.2 | NP_064418.1 | 1 | 730 | ||
GenBank mRNA | BC114579.1 | - | 1 | 658 |