siRNA Details
| Gene Symbol: Tmem9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020050.1 | NP_064434.1 | 5 | 1036 | ||
| GenBank mRNA | AK009147.1 | - | 5 | 971 | ||
| AK049882.1 | - | 5 | 1123 | |||
| AK077253.1 | - | 5 | 992 | |||
| AK147127.1 | - | 5 | 1052 | |||
| AK207353.1 | - | 1 | 367 | |||
| BC002208.1 | - | 5 | 1020 | 3490055 | ||

Tmem9b
Genomic Map