siRNA Details
| Gene Symbol: MRC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006039.4 | NP_006030.2 | 19 | 3179 | ||
| XM_011525543.2 | XP_011523845.1 | 19 | 2910 | |||
| GenBank mRNA | AB014609.2 | - | 19 | 3179 | ||
| AF107292.1 | - | 19 | 2820 | |||
| AF134838.1 | - | 19 | 2901 | |||
| AK093230.1 | - | 1 | 454 | |||
| BC146647.1 | - | 19 | 2898 | 40147137 | ||
| BC150212.1 | - | 19 | 2883 | 40148946 | ||
| BC153884.1 | - | 19 | 3179 | 8860500 | ||

MRC2
Genomic Map