siRNA Details
Gene Symbol: Olfr690 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020290.2 | NP_064686.2 | 1 | 810 | ||
GenBank mRNA | BC137757.1 | - | 1 | 810 | 8860776 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr690 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020290.2 | NP_064686.2 | 1 | 810 | ||
GenBank mRNA | BC137757.1 | - | 1 | 810 | 8860776 |