siRNA Details
| Gene Symbol: Zfp276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020497.2 | NP_065243.2 | 9 | 1951 | ||
| GenBank mRNA | AF178935.1 | - | 8 | 993 | ||
| AK028939.1 | - | 9 | 1904 | |||
| AK133363.1 | - | 9 | 1952 | |||
| BC066056.1 | - | 8 | 1898 | |||
| BC078415.1 | - | 9 | 1335 | |||
| BC138579.1 | - | 8 | 943 | 8861599 | ||
| BC138580.1 | - | 8 | 943 | 8861600 | ||
| BC145339.1 | - | 7 | 979 | |||
| BC145340.1 | - | 8 | 943 | |||

Zfp276
Genomic Map