siRNA Details
| Gene Symbol: Sult3a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020565.2 | NP_065590.2 | 3 | 255 | ||
| GenBank mRNA | AF026075.1 | - | 3 | 256 | ||
| BC125593.1 | - | 3 | 255 | 40130108 | ||
| BC125595.1 | - | 3 | 255 | 40130110 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sult3a1| Gene Symbol: Sult3a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020565.2 | NP_065590.2 | 3 | 255 | ||
| GenBank mRNA | AF026075.1 | - | 3 | 256 | ||
| BC125593.1 | - | 3 | 255 | 40130108 | ||
| BC125595.1 | - | 3 | 255 | 40130110 | ||