siRNA Details
| Gene Symbol: Pth | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020623.2 | NP_065648.1 | 2, 3 | 222 | ||
| GenBank mRNA | AK138677.1 | - | 2, 3 | 220 | ||
| BC099456.1 | - | 2, 3 | 207 | 30845904 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pth| Gene Symbol: Pth | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020623.2 | NP_065648.1 | 2, 3 | 222 | ||
| GenBank mRNA | AK138677.1 | - | 2, 3 | 220 | ||
| BC099456.1 | - | 2, 3 | 207 | 30845904 | ||