siRNA Details
| Gene Symbol: Olfr1509 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020514.2 | NP_065260.2 | 1 | 447 | ||
| GenBank mRNA | BC119195.1 | - | 1 | 447 | 8733944 | |
| BC119199.1 | - | 1 | 447 | 8733948 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1509| Gene Symbol: Olfr1509 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020514.2 | NP_065260.2 | 1 | 447 | ||
| GenBank mRNA | BC119195.1 | - | 1 | 447 | 8733944 | |
| BC119199.1 | - | 1 | 447 | 8733948 | ||