siRNA Details
| Gene Symbol: Olfr140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020515.1 | NP_065261.1 | 1 | 266 | ||
| GenBank mRNA | BC111111.1 | - | 1 | 263 | ||
| BC141888.1 | - | 1 | 265 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr140| Gene Symbol: Olfr140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020515.1 | NP_065261.1 | 1 | 266 | ||
| GenBank mRNA | BC111111.1 | - | 1 | 263 | ||
| BC141888.1 | - | 1 | 265 | |||