siRNA Details
| Gene Symbol: Olfr17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020598.2 | NP_065623.2 | 1 | 275 | ||
| GenBank mRNA | BC139721.1 | - | 1 | 275 | 40042633 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr17| Gene Symbol: Olfr17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020598.2 | NP_065623.2 | 1 | 275 | ||
| GenBank mRNA | BC139721.1 | - | 1 | 275 | 40042633 | |