siRNA Details
| Gene Symbol: Wdr46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020603.2 | NP_065628.1 | 4, 5 | 485 | ||
| GenBank mRNA | AK145068.1 | - | 4, 5 | 540 | ||
| AK216977.1 | - | 2, 3 | 146 | |||
| BC013527.1 | - | 4, 5 | 528 | |||
| BC026431.1 | - | 1, 2 | 109 | |||
| BC046977.1 | - | 4, 5 | 485 | 6315172 | ||
| KJ650211.1 | - | 4, 5 | 470 | |||
| KJ663716.1 | - | 4, 5 | 470 | |||

Wdr46
Genomic Map