siRNA Details
| Gene Symbol: Ckb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021273.4 | NP_067248.1 | 6 | 873 | ||
| GenBank mRNA | AK002467.1 | - | 6 | 836 | ||
| AK014299.1 | - | 6 | 816 | |||
| AK148885.1 | - | 6 | 833 | |||
| AK152388.1 | - | 6 | 833 | |||
| AK153484.1 | - | 6 | 833 | |||
| AK161990.1 | - | 6 | 833 | |||
| AK165779.1 | - | 1 | 2120 | |||
| AK166980.1 | - | 6 | 836 | |||
| AK167034.1 | - | 6 | 836 | |||
| BC015271.1 | - | 6 | 803 | 4225384 | ||
| BC058257.1 | - | 1 | 2093 | |||
| BC106109.1 | - | 6 | 800 | 6395794 | ||

Ckb
Genomic Map