siRNA Details
| Gene Symbol: KCNK15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022358.3 | NP_071753.2 | 2 | 1137 | ||
| GenBank mRNA | AF257081.1 | - | 2 | 1137 | ||
| AF294350.1 | - | 2 | 1073 | |||
| AF294351.1 | - | 2 | 1073 | |||
| BC143282.1 | - | 2 | 1117 | 9051785 | ||

KCNK15
Genomic Map