siRNA Details
| Gene Symbol: Dnajc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020566.1 | NP_065591.1 | 4 | 473 | ||
| GenBank mRNA | AF036875.1 | - | 4 | 473 | ||
| AK008507.1 | - | 4 | 722 | |||
| AK170008.1 | - | 4 | 715 | |||
| BC002305.1 | - | 1 | 107 | |||
| BC049281.1 | - | 4 | 707 | 4989485 | ||

Dnajc4
Genomic Map