siRNA Details
| Gene Symbol: NUDT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001248011.1 | NP_001234940.1 | 2 | 552 | ||
| NM_024047.4 | NP_076952.1 | 2 | 552 | |||
| NM_198038.2 | NP_932155.1 | 2 | 194 | |||
| XM_011532031.2 | XP_011530333.1 | 2 | 555 | |||
| XM_011532032.2 | XP_011530334.1 | 2 | 2731 | |||
| GenBank mRNA | AF265341.1 | - | 2 | 193 | ||
| AF273028.1 | - | 2 | 516 | |||
| AK027295.1 | - | 2 | 542 | |||
| AK074845.1 | - | 2 | 194 | |||
| AK075408.1 | - | 2 | 472 | |||
| AK308028.1 | - | 2 | 319 | |||
| AY026252.1 | - | 2 | 547 | |||
| AY359123.1 | - | 2 | 246 | |||
| BC000542.2 | - | 2 | 312 | 3163944 | ||
| BI552154.1 | - | 192 | ||||
| BM544951.1 | - | 2 | 564 | |||
| BP277630.1 | - | 2 | 552 | |||

NUDT9
Genomic Map