siRNA Details
| Gene Symbol: Gabarapl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020590.4 | NP_065615.1 | 4 | 979 | ||
| GenBank mRNA | AB041648.1 | - | 4 | 1003 | ||
| AF180518.1 | - | 4 | 1006 | |||
| AK011975.1 | - | 4 | 961 | |||
| AK018687.1 | - | 4 | 980 | |||
| AK159015.1 | - | 4 | 974 | |||
| AK167500.1 | - | 4 | 972 | |||
| AK168921.1 | - | 4 | 1011 | |||
| BC004602.1 | - | 4 | 914 | 3585132 | ||
| BC024706.1 | - | 4 | 978 | 3711973 | ||

Gabarapl1
Genomic Map