siRNA Details
Gene Symbol: Olfr17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020598.2 | NP_065623.2 | 1 | 708 | ||
GenBank mRNA | BC139721.1 | - | 1 | 708 | 40042633 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020598.2 | NP_065623.2 | 1 | 708 | ||
GenBank mRNA | BC139721.1 | - | 1 | 708 | 40042633 |