siRNA Details
| Gene Symbol: Evc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021292.2 | NP_067267.2 | 7 | 906 | ||
| GenBank mRNA | AJ250841.1 | - | 7 | 886 | ||
| AK039759.1 | - | 7 | 876 | |||
| AK148262.1 | - | 7 | 879 | |||
| BC021392.1 | - | 7 | 895 | 5069005 | ||
| BC021464.1 | - | 7 | 895 | 5069004 | ||

Evc
Genomic Map