siRNA Details
| Gene Symbol: Lancl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190984.1 | NP_001177913.1 | 7, 8 | 960 | ||
| NM_001190985.1 | NP_001177914.1 | 6, 7 | 1178 | |||
| NM_021295.3 | NP_067270.1 | 7, 8 | 994 | |||
| GenBank mRNA | AJ294535.1 | - | 7, 8 | 987 | ||
| AK146120.1 | - | 6, 7 | 1169 | |||
| AK182820.1 | - | 1, 2 | 108 | |||
| BC058560.1 | - | 7, 8 | 960 | 6315377 | ||
| Y11550.1 | - | 7, 8 | 959 | |||
| Y16518.1 | - | 6, 7 | 1138 | |||

Lancl1
Genomic Map